accessibility menu, dialog, popup

UserName

CAS RN 827616-52-4

IUPAC Name: 3-iodopyridine-2-carbonitrile
Synonyms: 3-iodopyridine-2-carbonitrile
Molecular Weight (g/mol): 230.01
Molecular Formula: C6H3IN2
InChi Key: MZZGCCSFZCRVAF-UHFFFAOYSA-N
SMILES: IC1=C(N=CC=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results