Learn More
2-[cis-4-(Boc-amino)cyclohexyl]ethylamine, 97%
CAS: 509143-12-8 | C13H26N2O2 | 242.363 g/mol
Chemical Identifiers
| CAS | 509143-12-8 |
|---|---|
| Molecular Formula | C13H26N2O2 |
| Molecular Weight (g/mol) | 242.363 |
| MDL Number | MFCD03844601 |
| InChI Key | SSZKBGFWNFAXNC-UHFFFAOYSA-N |
| Synonym | tert-butyl 4-2-aminoethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino-cyclohexyl ethylamine, tert-butyl cis-4-2-aminoethyl cyclohexylcarbamate, carbamicacid, cis-4-2-aminoethyl cyclohexyl-, 1,1-dimethylethyl ester 9ci, tert-butyl n-4-2-aminoethyl cyclohexyl carbamate, tert-butyl n-4-2-azanylethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino cyclohexyl ethylamine, tert-butyl trans-4-2-aminoethyl cyclohexyl carbamate |
| PubChem CID | 24720893 |
| IUPAC Name | tert-butyl N-[4-(2-aminoethyl)cyclohexyl]carbamate |
| SMILES | CC(C)(C)OC(=O)NC1CCC(CC1)CCN |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH62917MD
|
N/A | 250 mg |
N/A
|
|
|||||
|
AAH6291703
|
N/A | 1 g |
N/A
|
|
|||||
Description
2-[cis-4-(Boc-amino)cyclohexyl]ethylamine is used as a intermediate for chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-[cis-4-(Boc-amino)cyclohexyl]ethylamine is used as a intermediate for chemical research.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
| 509143-12-8 | |
| 242.363 | |
| SSZKBGFWNFAXNC-UHFFFAOYSA-N | |
| 24720893 | |
| CC(C)(C)OC(=O)NC1CCC(CC1)CCN |
| C13H26N2O2 | |
| MFCD03844601 | |
| tert-butyl 4-2-aminoethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino-cyclohexyl ethylamine, tert-butyl cis-4-2-aminoethyl cyclohexylcarbamate, carbamicacid, cis-4-2-aminoethyl cyclohexyl-, 1,1-dimethylethyl ester 9ci, tert-butyl n-4-2-aminoethyl cyclohexyl carbamate, tert-butyl n-4-2-azanylethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino cyclohexyl ethylamine, tert-butyl trans-4-2-aminoethyl cyclohexyl carbamate | |
| tert-butyl N-[4-(2-aminoethyl)cyclohexyl]carbamate |
Specifications
| 509143-12-8 | |
| MFCD03844601 | |
| SSZKBGFWNFAXNC-UHFFFAOYSA-N | |
| tert-butyl N-[4-(2-aminoethyl)cyclohexyl]carbamate | |
| 24720893 | |
| 97% |
| C13H26N2O2 | |
| tert-butyl 4-2-aminoethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino-cyclohexyl ethylamine, tert-butyl cis-4-2-aminoethyl cyclohexylcarbamate, carbamicacid, cis-4-2-aminoethyl cyclohexyl-, 1,1-dimethylethyl ester 9ci, tert-butyl n-4-2-aminoethyl cyclohexyl carbamate, tert-butyl n-4-2-azanylethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino cyclohexyl ethylamine, tert-butyl trans-4-2-aminoethyl cyclohexyl carbamate | |
| CC(C)(C)OC(=O)NC1CCC(CC1)CCN | |
| 242.363 | |
| 242.35 | |
| 2-[cis-4-(Boc-amino)cyclohexyl]ethylamine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only