missing translation for 'onlineSavingsMsg'
Learn More

2-[cis-4-(Boc-amino)cyclohexyl]ethylamine, 97%

Catalog No. AAH62917MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62917MD 250 mg
AAH6291703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH62917MD Supplier Thermo Scientific Chemicals Supplier No. H62917MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 509143-12-8 | C13H26N2O2 | 242.363 g/mol

2-[cis-4-(Boc-amino)cyclohexyl]ethylamine is used as a intermediate for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-[cis-4-(Boc-amino)cyclohexyl]ethylamine is used as a intermediate for chemical research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 509143-12-8
Molecular Formula C13H26N2O2
Molecular Weight (g/mol) 242.363
MDL Number MFCD03844601
InChI Key SSZKBGFWNFAXNC-UHFFFAOYSA-N
Synonym tert-butyl 4-2-aminoethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino-cyclohexyl ethylamine, tert-butyl cis-4-2-aminoethyl cyclohexylcarbamate, carbamicacid, cis-4-2-aminoethyl cyclohexyl-, 1,1-dimethylethyl ester 9ci, tert-butyl n-4-2-aminoethyl cyclohexyl carbamate, tert-butyl n-4-2-azanylethyl cyclohexyl carbamate, 2-trans-4-tert-butoxycarbonylamino cyclohexyl ethylamine, tert-butyl trans-4-2-aminoethyl cyclohexyl carbamate
PubChem CID 24720893
IUPAC Name tert-butyl N-[4-(2-aminoethyl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)CCN

Specifications

Quantity 250 mg
Formula Weight 242.35
Percent Purity 97%
Chemical Name or Material 2-[cis-4-(Boc-amino)cyclohexyl]ethylamine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only