missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorothioxanthone 98.0+%, TCI America™
Supplier: TCI America C029225G
Specifications
| 2-Chlorothioxanthone | |
| 152°C | |
| MFCD00005067 | |
| 1325 | |
| ZCDADJXRUCOCJE-UHFFFAOYSA-N | |
| 2-chloro-9H-thioxanthen-9-one | |
| 618848 | |
| ≥98.0% (GC) |
| 86-39-5 | |
| C13H7ClOS | |
| 25 g | |
| 2-chlorothioxanthone, 2-chloro-9h-thioxanthen-9-one, 9h-thioxanthen-9-one, 2-chloro, unii-rgs87t004b, 2-chlorothiaxanthone, 2-chlorothioxanthene-9-one, sandoray 1050, 2-chloro-thioxanthone, pubchem10692, 2-chloro-9-thioxanthone | |
| ClC1=CC=C2SC3=CC=CC=C3C(=O)C2=C1 | |
| 246.71 | |
| 246.71 | |
| Crystalline Powder |
Chemical Identifiers
| 86-39-5 | |
| 246.71 | |
| ZCDADJXRUCOCJE-UHFFFAOYSA-N | |
| 618848 | |
| ClC1=CC=C2SC3=CC=CC=C3C(=O)C2=C1 |
| C13H7ClOS | |
| MFCD00005067 | |
| 2-chlorothioxanthone, 2-chloro-9h-thioxanthen-9-one, 9h-thioxanthen-9-one, 2-chloro, unii-rgs87t004b, 2-chlorothiaxanthone, 2-chlorothioxanthene-9-one, sandoray 1050, 2-chloro-thioxanthone, pubchem10692, 2-chloro-9-thioxanthone | |
| 2-chloro-9H-thioxanthen-9-one |
Safety and Handling
EINECSNumber : (5)-5537
TSCA : Yes