missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothiophenol, 98%

Catalog No. p-7081846
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1201414 25 g
AAA1201430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1201414 Supplier Thermo Scientific Chemicals Supplier No. A1201414
Only null left

CAS: 6320-03-2 | C6H5ClS | 144.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6320-03-2
Molecular Formula C6H5ClS
Molecular Weight (g/mol) 144.62
MDL Number MFCD00004830
InChI Key PWOBDMNCYMQTCE-UHFFFAOYSA-N
Synonym 2-chlorothiophenol, o-chlorothiophenol, benzenethiol, 2-chloro, 1-chloro-2-mercaptobenzene, o-chlorobenzenethiol, o-chloromercaptobenzene, chlorothiophenol, benzenethiol, o-chloro, 2-chlorobenzene-1-thiol, 2-chloro thiophenol
PubChem CID 80599
IUPAC Name 2-chlorobenzenethiol
SMILES C1=CC=C(C(=C1)S)Cl

Specifications

Density 1.277
Boiling Point 108°C to 110°C (30 mmHg)
Flash Point 88°C (190°F)
Refractive Index 1.604
Quantity 25 g
UN Number UN1760
Beilstein 1209665
Sensitivity Air Sensitive
Formula Weight 144.62
Percent Purity 98%
Chemical Name or Material 2-Chlorothiophenol
Show More Show Less
Hazard Category H227-H302-H314-H335
Hazard Statement GHS H Statement
H301-H314-H318-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 228-667-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only