missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorostyrene, 95%, stab. with 0.1% 4-tert-butylcatechol

Catalog No. p-7025849
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2155106 5 g
AAB2155114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2155106 Supplier Thermo Scientific Chemicals Supplier No. B2155106
Only null left

CAS: 2039-87-4 | C8H7Cl | 138.594 g/mol

2-Chlorostyrene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorostyrene is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.

Chemical Identifiers

CAS 2039-87-4
Molecular Formula C8H7Cl
Molecular Weight (g/mol) 138.594
MDL Number MFCD00000567
InChI Key ISRGONDNXBCDBM-UHFFFAOYSA-N
Synonym 2-chlorostyrene, 1-chloro-2-vinylbenzene, o-chlorostyrene, benzene, 1-chloro-2-ethenyl, styrene, o-chloro, ortho-chlorostyrene, stryene, o-chloro, styrene, 2-chloro, unii-za43r4q315, ccris 4598
PubChem CID 14906
IUPAC Name 1-chloro-2-ethenylbenzene
SMILES C=CC1=CC=CC=C1Cl

Specifications

Melting Point -63°C
Density 1.091
Boiling Point 189°C
Flash Point 65°C (149°F)
Odor Pungent
Refractive Index 1.565
Quantity 5 g
Beilstein 2038863
Solubility Information Insoluble in water.
Formula Weight 138.6
Percent Purity 98%
Chemical Name or Material 2-Chlorostyrene, stabilized with 0.1% 4-tert-butylcatechol
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 218-026-8
RTECSNumber WL4160000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only