missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrimidine 99.0+%, TCI America™

Catalog No. C105025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C105025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C105025G Supplier TCI America Supplier No. C105025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1722-12-9
Molecular Formula C4H3ClN2
Molecular Weight (g/mol) 114.53
MDL Number MFCD00006060
InChI Key UNCQVRBWJWWJBF-UHFFFAOYSA-N
Synonym pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin
PubChem CID 74404
IUPAC Name 2-chloropyrimidine
SMILES ClC1=NC=CC=N1

Specifications

Melting Point 63°C
Color White-Yellow
Boiling Point 91°C
Quantity 25 g
Formula Weight 114.53
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloropyrimidine