accessibility menu, dialog, popup

UserName

ClNN

CAS RN 1722-12-9

IUPAC Name: 2-chloropyrimidine
Synonyms: pyrimidine, 2-chloro-
Molecular Weight (g/mol): 114.53
Molecular Formula: C4H3ClN2
InChi Key: UNCQVRBWJWWJBF-UHFFFAOYSA-N
SMILES: ClC1=NC=CC=N1
Boiling Point 
  • (3)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)