missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrimidine, 98+%

CAS: 1722-12-9 | C4H3ClN2 | 114.53 g/mol

$136.72 - $136.72

Chemical Identifiers

CAS 1722-12-9
Molecular Formula C4H3ClN2
Molecular Weight (g/mol) 114.53
MDL Number MFCD00006060
InChI Key UNCQVRBWJWWJBF-UHFFFAOYSA-N
Synonym pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin
PubChem CID 74404
IUPAC Name 2-chloropyrimidine
SMILES ClC1=NC=CC=N1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC185840100
SDS View Documents
Thermo Scientific Chemicals
185840100
10 g Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1722-12-9
114.53
UNCQVRBWJWWJBF-UHFFFAOYSA-N
74404
ClC1=NC=CC=N1
C4H3ClN2
MFCD00006060
pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin
2-chloropyrimidine

Specifications

1722-12-9
Orange to Yellow
98°C
98% min. (GC)
C4H3ClN2
10 g
UNCQVRBWJWWJBF-UHFFFAOYSA-N
2-chloropyrimidine
74404
98+%
2-Chloropyrimidine
62.0°C to 66.0°C
75.0°C to 76.0°C (10.0 mmHg)
Authentic
Glass bottle
MFCD00006060
pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin
ClC1=NC=CC=N1
114.53
114.53
Crystalline Powder or Crystals
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 217-020-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only