Learn More
2-Chloropyrimidine, 98%
CAS: 1722-12-9 | C4H3ClN2 | 114.53 g/mol
Supplier: Thermo Scientific Chemicals A1539414
Description
2-Chloropyrimidine is a useful reagent in the preparation of antivirals and other biologically active compounds. It was used in the synthesis of 4?-(1,1?-(5-(2-methoxyphenoxy)-[2,2?-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay, 2-amino-4-heteroarylpyrimidines, cis- and trans-octahydropyrrolo[2,3]pyridine derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloropyrimidine is a useful reagent in the preparation of antivirals and other biologically active compounds. It was used in the synthesis of 4′-(1,1′-(5-(2-methoxyphenoxy)-[2,2′-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay, 2-amino-4-heteroarylpyrimidines, cis- and trans-octahydropyrrolo[2,3]pyridine derivatives.
Solubility
Slightly soluble in water. 33.3 mg soluble in 1 mL of alcohol.
Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents. Store in refrigerator.
Specifications
| 2-Chloropyrimidine | |
| 64°C to 67°C | |
| 98°C (208°F) | |
| MFCD00006060 | |
| UN2811 | |
| Hygroscopic | |
| Slightly soluble in water. 33.3 mg soluble in 1 mL of alcohol. | |
| ClC1=NC=CC=N1 | |
| 114.53 | |
| 114.54 |
| 1722-12-9 | |
| 75°C to 76°C (10 mmHg) | |
| C4H3ClN2 | |
| 25 g | |
| 107171 | |
| pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin | |
| UNCQVRBWJWWJBF-UHFFFAOYSA-N | |
| 2-chloropyrimidine | |
| 74404 | |
| 98% |
Chemical Identifiers
| 1722-12-9 | |
| 114.53 | |
| UNCQVRBWJWWJBF-UHFFFAOYSA-N | |
| 74404 | |
| ClC1=NC=CC=N1 |
| C4H3ClN2 | |
| MFCD00006060 | |
| pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin | |
| 2-chloropyrimidine |
Safety and Handling
GHS H Statement
H302-H319-H317-H335
Harmful if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 217-020-2
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only