missing translation for 'onlineSavingsMsg'
Learn More

2-chloropyrimidine-5-carboxaldehyde, 97%

Catalog No. AC438450050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438450050 5 g Glass bottle
AC438450010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438450050 Supplier Thermo Scientific Chemicals Supplier No. 438450050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 933702-55-7 | C5H3ClN2O | 142.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 933702-55-7
Molecular Formula C5H3ClN2O
Molecular Weight (g/mol) 142.54
MDL Number MFCD10696891
InChI Key LJYQVOPFBNMTKJ-UHFFFAOYSA-N
Synonym 2-chloropyrimidine-5-carboxyaldehyde, 2-chloropyrimidine-5-carboxaldehyde, 2-chloro-5-pyrimidinecarboxaldehyde, pubchem23896, 2-chloro-5-formyl pyrimidine, chloropyrimidine-5-carbaldehyde, 2-chloro-pyrimidine-5-carbaldehyde, 2-chloranylpyrimidine-5-carbaldehyde, 5-pyrimidinecarboxaldehyde, 2-chloro
PubChem CID 21698350
IUPAC Name 2-chloropyrimidine-5-carbaldehyde
SMILES ClC1=NC=C(C=O)C=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 113°C to 118°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 142.55
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Chloropyrimidine-5-carboxaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

RUO – Research Use Only