accessibility menu, dialog, popup

UserName

NNClO

CAS RN 933702-55-7

IUPAC Name: 2-chloropyrimidine-5-carbaldehyde
Synonyms: 2-chloropyrimidine-5-carbaldehyde
Molecular Weight (g/mol): 142.54
Molecular Formula: C5H3ClN2O
InChi Key: LJYQVOPFBNMTKJ-UHFFFAOYSA-N
SMILES: ClC1=NC=C(C=O)C=N1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results