missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrimidine-5-boronic acid, 96%

Catalog No. AAH6494903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6494903 1 g
AAH6494906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6494903 Supplier Thermo Scientific Chemicals Supplier No. H6494903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1003845-06-4 | C4H4BClN2O2 | 158.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1003845-06-4
Molecular Formula C4H4BClN2O2
Molecular Weight (g/mol) 158.35
MDL Number MFCD08063113
InChI Key YTCIHPTZKKWKKC-UHFFFAOYSA-N
Synonym 2-chloropyrimidine-5-boronic acid, 2-chloro-5-pyrimidineboronic acid, 2-chloropyrimidin-5-yl boronic acid, acmc-2097pq, ksc493q8p, 2-chloropyrimidin-5-boronic acid, 2-chloro-5-pyrimidinylboronic acid, 2-chloro-pyridineyl-5-boronic acid, 2-chloro-pyrimidine-5-boronic acid
PubChem CID 21747362
IUPAC Name (2-chloropyrimidin-5-yl)boronic acid
SMILES OB(O)C1=CN=C(Cl)N=C1

Specifications

Color White
Quantity 1 g
Formula Weight 158.35
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 2-Chloropyrimidine-5-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only