missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloropropionic Acid 98.0+%, TCI America™
$78.29 - $274.02
Chemical Identifiers
| CAS | 598-78-7 |
|---|---|
| Molecular Formula | C3H5ClO2 |
| Molecular Weight (g/mol) | 108.52 |
| MDL Number | MFCD00004224 |
| InChI Key | GAWAYYRQGQZKCR-UHFFFAOYNA-N |
| Synonym | 2-chloropropionic acid, propanoic acid, 2-chloro, propionic acid, 2-chloro, alpha-chloropropionic acid, alpha-monochloropropionic acid, propionic acid, alpha-chloro, .alpha.-chloropropionic acid, propionic acid, .alpha.-chloro, chloropropionic acid, propanoic acid, chloro |
| PubChem CID | 11734 |
| IUPAC Name | 2-chloropropanoic acid |
| SMILES | CC(Cl)C(O)=O |
Chemical Identifiers
| 598-78-7 | |
| 108.52 | |
| GAWAYYRQGQZKCR-UHFFFAOYNA-N | |
| 11734 | |
| CC(Cl)C(O)=O |
| C3H5ClO2 | |
| MFCD00004224 | |
| 2-chloropropionic acid, propanoic acid, 2-chloro, propionic acid, 2-chloro, alpha-chloropropionic acid, alpha-monochloropropionic acid, propionic acid, alpha-chloro, .alpha.-chloropropionic acid, propionic acid, .alpha.-chloro, chloropropionic acid, propanoic acid, chloro | |
| 2-chloropropanoic acid |
Specifications
| 598-78-7 | |
| Yellow | |
| C3H5ClO2 | |
| 25 g | |
| 2-chloropropionic acid, propanoic acid, 2-chloro, propionic acid, 2-chloro, alpha-chloropropionic acid, alpha-monochloropropionic acid, propionic acid, alpha-chloro, .alpha.-chloropropionic acid, propionic acid, .alpha.-chloro, chloropropionic acid, propanoic acid, chloro | |
| CC(Cl)C(O)=O | |
| 108.52 | |
| 108.52 | |
| Liquid |
| -12°C | |
| 186°C | |
| MFCD00004224 | |
| 2511 | |
| GAWAYYRQGQZKCR-UHFFFAOYNA-N | |
| 2-chloropropanoic acid | |
| 11734 | |
| ≥98.0% (GC,T) | |
| 2-Chloropropionic Acid |
Safety and Handling
EINECSNumber : (2)-1157
RTECSNumber : UE8575000
TSCA : Yes