Learn More
2-Chlorophenylhydrazine hydrochloride, 97%
Description
2-Chlorophenylhydrazine hydrochloride is used to study intrinsic resistance of Mycobacterium smegmatis to fluoroquinolones, used for treatment of Mycobacterium tuberculosis. It may be used in pyrazoline synthesis. It can be used to produce N-azepan-2-ylidene-N'-(2-chloro-phenyl)-hydrazine with 7-methoxy-3,4,5,6-tetrahydro-2H-azepine at heating.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Chlorophenylhydrazine hydrochloride is used to study intrinsic resistance of Mycobacterium smegmatis to fluoroquinolones, used for treatment of Mycobacterium tuberculosis. It may be used in pyrazoline synthesis. It can be used to produce N-azepan-2-ylidene-N′-(2-chloro-phenyl)-hydrazine with 7-methoxy-3,4,5,6-tetrahydro-2H-azepine at heating.
Solubility
Soluble in water.
Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
Specifications
Specifications
| Melting Point | ∼201°C (decomposition) |
| Quantity | 100 g |
| Beilstein | 3699381 |
| Solubility Information | Soluble in water. |
| Formula Weight | 179.05 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Chlorophenylhydrazine hydrochloride |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H301-H315-H319-H335 Toxic if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 255-194-1 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only