missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl Isothiocyanate 95.0+%, TCI America™

Catalog No. I05345G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05345G 5 g
I053425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05345G Supplier TCI America Supplier No. I05345G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2740-81-0
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
MDL Number MFCD00004801
InChI Key DASSPOJBUMBXLU-UHFFFAOYSA-N
Synonym 2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, 1-chloro-2-isothiocyanato-benzene, isothiocyanic acid 2-chlorophenyl ester, 2-chlorobenzenisothiocyanate, acmc-1ccw5, o-chlorophenylisothiocyanate, o-chlorophenyl isothiocyanate
PubChem CID 123171
IUPAC Name 1-chloro-2-isothiocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=S)Cl

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 2810
Boiling Point 262°C
Formula Weight 169.63
Percent Purity ≥95.0% (GC)
Chemical Name or Material 2-Chlorophenyl Isothiocyanate