accessibility menu, dialog, popup

UserName

CAS RN 2740-81-0

IUPAC Name: 1-chloro-2-isothiocyanatobenzene
Synonyms: 1-chloro-2-isothiocyanatobenzene
Molecular Weight (g/mol): 169.63
Molecular Formula: C7H4ClNS
InChi Key: DASSPOJBUMBXLU-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1N=C=S
Molecular Weight (g/mol) 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (3)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)