missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl isothiocyanate, 99%, Thermo Scientific™

Chemical Identifiers

CAS 2740-81-0
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.63
MDL Number MFCD00004801
InChI Key DASSPOJBUMBXLU-UHFFFAOYSA-N
Synonym 2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, 1-chloro-2-isothiocyanato-benzene, isothiocyanic acid 2-chlorophenyl ester, 2-chlorobenzenisothiocyanate, acmc-1ccw5, o-chlorophenylisothiocyanate, o-chlorophenyl isothiocyanate
PubChem CID 123171
IUPAC Name 1-chloro-2-isothiocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=S)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC292240050
SDS View Documents
Thermo Scientific Chemicals
292240050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2740-81-0
169.63
DASSPOJBUMBXLU-UHFFFAOYSA-N
123171
C1=CC=C(C(=C1)N=C=S)Cl
C7H4ClNS
MFCD00004801
2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, 1-chloro-2-isothiocyanato-benzene, isothiocyanic acid 2-chlorophenyl ester, 2-chlorobenzenisothiocyanate, acmc-1ccw5, o-chlorophenylisothiocyanate, o-chlorophenyl isothiocyanate
1-chloro-2-isothiocyanatobenzene

Specifications

2740-81-0
100.0
260°C to 262°C
Authentic
Glass bottle
1.6598 to 1.6618
MFCD00004801
12, 601
2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, 1-chloro-2-isothiocyanato-benzene, isothiocyanic acid 2-chlorophenyl ester, 2-chlorobenzenisothiocyanate, acmc-1ccw5, o-chlorophenylisothiocyanate, o-chlorophenyl isothiocyanate
C1=CC=C(C(=C1)N=C=S)Cl
169.63
169.63
2-Chlorophenyl isothiocyanate, 99%
98.5
1.2900g/mL
>110°C
98.5% min. (GC)
C7H4ClNS
NCSC6H4Cl
5g
1.29
DASSPOJBUMBXLU-UHFFFAOYSA-N
1-chloro-2-isothiocyanatobenzene
123171
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash

GHS Signal Word: Danger

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only