missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenyl isothiocyanate, 97%

Catalog No. AAA1823806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1823806 5 g
AAA1823814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1823806 Supplier Thermo Scientific Chemicals Supplier No. A1823806

CAS: 2740-81-0 | C7H4ClNS | 169.626 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2740-81-0
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.626
MDL Number MFCD00004801
InChI Key DASSPOJBUMBXLU-UHFFFAOYSA-N
Synonym 2-chlorophenyl isothiocyanate, 2-chlorophenylisothiocyanate, benzene, 1-chloro-2-isothiocyanato, benzene, chloroisothiocyanato, 1-chloro-2-isothiocyanato-benzene, isothiocyanic acid 2-chlorophenyl ester, 2-chlorobenzenisothiocyanate, acmc-1ccw5, o-chlorophenylisothiocyanate, o-chlorophenyl isothiocyanate
PubChem CID 123171
IUPAC Name 1-chloro-2-isothiocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=S)Cl

Specifications

Density 1.287
Quantity 5 g
Odor Pungent
Refractive Index 1.6608
UN Number UN2922
Beilstein 2081473
Boiling Point 260°C to 262°C
Formula Weight 169.64
Percent Purity 97%
Chemical Name or Material 2-Chlorophenyl isothiocyanate
Hazard Category H301+H311+H331-H315-H319-H334-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P261-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only