missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenoxyacetic acid, 98+%

CAS: 614-61-9 | C8H7ClO3 | 186.59 g/mol

Supplier:  Thermo Scientific Chemicals L0092622

 View more versions of this product

Catalog No. AAL0092622


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chlorophenoxyacetic acid
614-61-9
C8H7ClO3
1103151
OPQYFNRLWBWCST-UHFFFAOYSA-N
2-(2-chlorophenoxy)acetic acid
11969
≥98%
147°C to 152°C
100 g
MFCD00004298
2-chlorophenoxyacetic acid, 2-chlorophenoxy acetic acid, o-chlorophenoxyacetic acid, 2-2-chlorophenoxy acetic acid, acetic acid, 2-chlorophenoxy, acide o-chlorophenoxyacetique, acetic acid, o-chlorophenoxy, unii-x2512m5l7q, acetic acid, chlorophenoxy
OC(=O)COC1=CC=CC=C1Cl
186.59
186.59

Chemical Identifiers

614-61-9
186.59
OPQYFNRLWBWCST-UHFFFAOYSA-N
11969
OC(=O)COC1=CC=CC=C1Cl
C8H7ClO3
MFCD00004298
2-chlorophenoxyacetic acid, 2-chlorophenoxy acetic acid, o-chlorophenoxyacetic acid, 2-2-chlorophenoxy acetic acid, acetic acid, 2-chlorophenoxy, acide o-chlorophenoxyacetique, acetic acid, o-chlorophenoxy, unii-x2512m5l7q, acetic acid, chlorophenoxy
2-(2-chlorophenoxy)acetic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 210-388-5

RTECSNumber : AF9900000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only