accessibility menu, dialog, popup

UserName

HOOOCl

CAS RN 614-61-9

IUPAC Name: 2-(2-chlorophenoxy)acetic acid
Synonyms: O-chlorophenoxyacetic acid
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: OPQYFNRLWBWCST-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=CC=C1Cl
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)