missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenoxyacetic acid, 98+%

Catalog No. AAL0092614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0092614 25 g
AAL0092622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0092614 Supplier Thermo Scientific Chemicals Supplier No. L0092614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 614-61-9 | C8H7ClO3 | 186.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 614-61-9
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD00004298
InChI Key OPQYFNRLWBWCST-UHFFFAOYSA-N
Synonym 2-chlorophenoxyacetic acid, 2-chlorophenoxy acetic acid, o-chlorophenoxyacetic acid, 2-2-chlorophenoxy acetic acid, acetic acid, 2-chlorophenoxy, acide o-chlorophenoxyacetique, acetic acid, o-chlorophenoxy, unii-x2512m5l7q, acetic acid, chlorophenoxy
PubChem CID 11969
IUPAC Name 2-(2-chlorophenoxy)acetic acid
SMILES OC(=O)COC1=CC=CC=C1Cl

Specifications

Melting Point 147°C to 152°C
Quantity 25 g
Beilstein 1103151
Formula Weight 186.59
Percent Purity ≥98%
Chemical Name or Material 2-Chlorophenoxyacetic acid
Hazard Category H302
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 210-388-5
RTECSNumber AF9900000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only