missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorophenothiazine 98.0+%, TCI America™

Catalog No. C024825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C024825G 25 g
C0248500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C024825G Supplier TCI America Supplier No. C024825G

Chemical Identifiers

CAS 92-39-7
Molecular Formula C12H8ClNS
Molecular Weight (g/mol) 233.713
MDL Number MFCD00005016
InChI Key KFZGLJSYQXZIGP-UHFFFAOYSA-N
Synonym 2-chlorophenothiazine, 10h-phenothiazine, 2-chloro, phenothiazine, 2-chloro, 2-chlorothiodiphenylamine, wln: t c666 bm isj eg, 2-chloro-phenothiazine, 2-chloropheno-thiazine, acmc-209rfl, 2-chlorophenothizine, dsstox_cid_31643
PubChem CID 7088
IUPAC Name 2-chloro-10H-phenothiazine
SMILES C1=CC=C2C(=C1)NC3=C(S2)C=CC(=C3)Cl

Specifications

Melting Point 198°C
Quantity 25 g
Formula Weight 233.71
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorophenothiazine
EINECSNumber (5)-3803
RTECSNumber SN7580000
TSCA Yes