missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronitrobenzene 99.0+%, TCI America™

Catalog No. p-7124289
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C022025G 25 g
C0220500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C022025G Supplier TCI America Supplier No. C022025G

Chemical Identifiers

CAS 88-73-3
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.55
MDL Number MFCD00007061
InChI Key BFCFYVKQTRLZHA-UHFFFAOYSA-N
Synonym 2-chloronitrobenzene, o-chloronitrobenzene, 2-nitrochlorobenzene, chloronitrobenzene, o-nitrochlorobenzene, benzene, 1-chloro-2-nitro, 2-chloro-1-nitrobenzene, oncb, benzene, chloronitro, nitrochlorobenzene
PubChem CID 6945
ChEBI CHEBI:34270
IUPAC Name 1-chloro-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC=CC=C1Cl

Specifications

Color Yellow
Boiling Point 246°C
Quantity 25 g
UN Number 1578
Formula Weight 157.55
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Chloronitrobenzene
EINECSNumber (3)-0442
RTECSNumber CZ0875000
TSCA Yes