missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronicotinoyl chloride, 97%

Catalog No. p-4089962
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC395790050 5 g Glass bottle
AC395790250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC395790050 Supplier Thermo Scientific Chemicals Supplier No. 395790050

CAS: 49609-84-9 | C6H3Cl2NO | 176.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 49609-84-9
Molecular Formula C6H3Cl2NO
Molecular Weight (g/mol) 176.00
MDL Number MFCD00051677
InChI Key RXTRRIFWCJEMEL-UHFFFAOYSA-N
Synonym 2-chloronicotinoyl chloride, 2-chloro-nicotinoyl chloride, 3-pyridinecarbonyl chloride, 2-chloro, 2-chloronicotinylchloride, pubchem2599, acmc-209kgw, chloronicotinoyl chloride, 2-chloronicotinoylchloride, 2-chloro nicotinoyl chloride, 2-chloronicotinic acid chloride
PubChem CID 2774541
IUPAC Name 2-chloropyridine-3-carbonyl chloride
SMILES ClC(=O)C1=CC=CN=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 38°C to 39°C
Color Beige-Yellow to White
Boiling Point 89°C to 91°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 5 g
Formula Weight 176
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 2-Chloronicotinoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only