missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronicotinic Acid 98.0+%, TCI America™

$39.14 - $184.54

Chemical Identifiers

CAS 2942-59-8
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD00006236
InChI Key IBRSSZOHCGUTHI-UHFFFAOYSA-N
Synonym 2-chloronicotinic acid, 2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-chloro, 2-chloro-nicotinic acid, 2-chloro nicotinic acid, chloronicotinic acid, nicotinic acid, 2-chloro, 2-chloronicotinicacid, 2-chloroisonicotinicacid, 2chloronicotinic acid
PubChem CID 76258
IUPAC Name 2-chloropyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)Cl)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C110825G
View Documents
TCI America
C110825G
25 g N/A
Only null left
 
C1108250G
View Documents
TCI America
C1108250G
250 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2942-59-8
157.553
IBRSSZOHCGUTHI-UHFFFAOYSA-N
76258
C1=CC(=C(N=C1)Cl)C(=O)O
C6H4ClNO2
MFCD00006236
2-chloronicotinic acid, 2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-chloro, 2-chloro-nicotinic acid, 2-chloro nicotinic acid, chloronicotinic acid, nicotinic acid, 2-chloro, 2-chloronicotinicacid, 2-chloroisonicotinicacid, 2chloronicotinic acid
2-chloropyridine-3-carboxylic acid

Specifications

2942-59-8
MFCD00006236
2-chloronicotinic acid, 2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-chloro, 2-chloro-nicotinic acid, 2-chloro nicotinic acid, chloronicotinic acid, nicotinic acid, 2-chloro, 2-chloronicotinicacid, 2-chloroisonicotinicacid, 2chloronicotinic acid
C1=CC(=C(N=C1)Cl)C(=O)O
157.553
157.55
Crystalline Powder
C6H4ClNO2
25 g
IBRSSZOHCGUTHI-UHFFFAOYSA-N
2-chloropyridine-3-carboxylic acid
76258
≥98.0% (HPLC,T)
2-Chloronicotinic Acid
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-6195

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'