Learn More
2-Chloronicotinic acid, 99%
CAS: 2942-59-8 | C6H4ClNO2 | 157.553 g/mol
$53.48 - $713.18
Chemical Identifiers
| CAS | 2942-59-8 |
|---|---|
| Molecular Formula | C6H4ClNO2 |
| Molecular Weight (g/mol) | 157.553 |
| MDL Number | MFCD00006236 |
| InChI Key | IBRSSZOHCGUTHI-UHFFFAOYSA-N |
| Synonym | 2-chloronicotinic acid, 2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-chloro, 2-chloro-nicotinic acid, 2-chloro nicotinic acid, chloronicotinic acid, nicotinic acid, 2-chloro, 2-chloronicotinicacid, 2-chloroisonicotinicacid, 2chloronicotinic acid |
| PubChem CID | 76258 |
| IUPAC Name | 2-chloropyridine-3-carboxylic acid |
| SMILES | C1=CC(=C(N=C1)Cl)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1708209
|
Thermo Scientific Chemicals
A1708209 |
10 g |
N/A
|
|
|||||
|
AAA1708218
|
Thermo Scientific Chemicals
A1708218 |
50 g |
N/A
|
|
|||||
|
AAA1708230
|
Thermo Scientific Chemicals
A1708230 |
250 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2942-59-8 | |
| 157.553 | |
| IBRSSZOHCGUTHI-UHFFFAOYSA-N | |
| 76258 | |
| C1=CC(=C(N=C1)Cl)C(=O)O |
| C6H4ClNO2 | |
| MFCD00006236 | |
| 2-chloronicotinic acid, 2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-chloro, 2-chloro-nicotinic acid, 2-chloro nicotinic acid, chloronicotinic acid, nicotinic acid, 2-chloro, 2-chloronicotinicacid, 2-chloroisonicotinicacid, 2chloronicotinic acid | |
| 2-chloropyridine-3-carboxylic acid |
Specifications
| 2942-59-8 | |
| C6H4ClNO2 | |
| 10 g | |
| 2-chloronicotinic acid, 2-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 2-chloro, 2-chloro-nicotinic acid, 2-chloro nicotinic acid, chloronicotinic acid, nicotinic acid, 2-chloro, 2-chloronicotinicacid, 2-chloroisonicotinicacid, 2chloronicotinic acid | |
| C1=CC(=C(N=C1)Cl)C(=O)O | |
| 157.553 | |
| 157.56 | |
| 2-Chloronicotinic acid |
| ∼180°C (decomposition) | |
| MFCD00006236 | |
| 119023 | |
| IBRSSZOHCGUTHI-UHFFFAOYSA-N | |
| 2-chloropyridine-3-carboxylic acid | |
| 76258 | |
| 99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 220-937-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only