missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronaphthalene, For Spectrophotometric Det. of Proline and Thiophene, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111015903
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015903 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015903 Supplier Merck Emd Millipore Supplier No. 48517

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Environmental

Specifications

Packaging Ampule
CAS 91-58-7
Physical Form Neat
Linear Formula C10H7 Cl
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 162.62
Percent Purity ≥99.0%