missing translation for 'onlineSavingsMsg'
Learn More

2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 98%

CAS: 86604-75-3 | C9H13Cl2NO | 222.109 g/mol

Supplier:  Thermo Scientific Chemicals L1945314

 View more versions of this product

Catalog No. AAL1945314


Only null left
Explore available promotions

Description

Description

2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
Specifications

Specifications

2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride
∽128°C (decomposition)
MFCD00277472
5359162
Soluble in water.
CC1=CN=C(C(=C1OC)C)CCl.Cl
222.109
222.12
86604-75-3
C9H13Cl2NO
25 g
2-chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 2-chloromethyl-3,5-dimethyl-4-methoxypyridine hydrochloride, 2-chloromethyl-3,5-dimethyl-4-methoxy pyridine hydrochloride, 2-chloromethyl-3,5-dimethyl-4 methoxypyridine hydrochloride, 2-chloromethyl-4-methoxy-3,5-dimethylpyridine hcl, pyridine, 2-chloromethyl-4-methoxy-3,5-dimethyl-, hydrochloride, 2-chloromethyl-3,5-dimethyl-4-pyridinyl methyl ether hydrochloride, pubchem2236
LCJDHJOUOJSJGS-UHFFFAOYSA-N
2-(chloromethyl)-4-methoxy-3,5-dimethylpyridine;hydrochloride
11694258
98%

Chemical Identifiers

86604-75-3
222.109
LCJDHJOUOJSJGS-UHFFFAOYSA-N
11694258
CC1=CN=C(C(=C1OC)C)CCl.Cl
C9H13Cl2NO
MFCD00277472
2-chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 2-chloromethyl-3,5-dimethyl-4-methoxypyridine hydrochloride, 2-chloromethyl-3,5-dimethyl-4-methoxy pyridine hydrochloride, 2-chloromethyl-3,5-dimethyl-4 methoxypyridine hydrochloride, 2-chloromethyl-4-methoxy-3,5-dimethylpyridine hcl, pyridine, 2-chloromethyl-4-methoxy-3,5-dimethyl-, hydrochloride, 2-chloromethyl-3,5-dimethyl-4-pyridinyl methyl ether hydrochloride, pubchem2236
2-(chloromethyl)-4-methoxy-3,5-dimethylpyridine;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only