missing translation for 'onlineSavingsMsg'
Learn More

2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 98%

Catalog No. p-7047566
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1945306 5 g
AAL1945314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1945306 Supplier Thermo Scientific Chemicals Supplier No. L1945306
Only null left

CAS: 86604-75-3 | C9H13Cl2NO | 222.109 g/mol

2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride is used as pharmaceutical intermediates.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.

Chemical Identifiers

CAS 86604-75-3
Molecular Formula C9H13Cl2NO
Molecular Weight (g/mol) 222.109
MDL Number MFCD00277472
InChI Key LCJDHJOUOJSJGS-UHFFFAOYSA-N
Synonym 2-chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride, 2-chloromethyl-3,5-dimethyl-4-methoxypyridine hydrochloride, 2-chloromethyl-3,5-dimethyl-4-methoxy pyridine hydrochloride, 2-chloromethyl-3,5-dimethyl-4 methoxypyridine hydrochloride, 2-chloromethyl-4-methoxy-3,5-dimethylpyridine hcl, pyridine, 2-chloromethyl-4-methoxy-3,5-dimethyl-, hydrochloride, 2-chloromethyl-3,5-dimethyl-4-pyridinyl methyl ether hydrochloride, pubchem2236
PubChem CID 11694258
IUPAC Name 2-(chloromethyl)-4-methoxy-3,5-dimethylpyridine;hydrochloride
SMILES CC1=CN=C(C(=C1OC)C)CCl.Cl

Specifications

Melting Point ∽128°C (decomposition)
Quantity 5 g
Beilstein 5359162
Solubility Information Soluble in water.
Formula Weight 222.12
Percent Purity 98%
Chemical Name or Material 2-Chloromethyl-4-methoxy-3,5-dimethylpyridine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only