missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-3,4-dimethoxypyridine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7125270
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
C20735G 5 g
C207325G 25 g
2 options

Catalog No. C20735G

Supplier: TCI America C20735G

Only null left

Chemical Identifiers

CAS 72830-09-2
Molecular Formula C8H11Cl2NO2
Molecular Weight (g/mol) 224.081
MDL Number MFCD02181083
InChI Key YYRIKJFWBIEEDH-UHFFFAOYSA-N
Synonym 2-chloromethyl-3,4-dimethoxypyridine hydrochloride, 2-chloromethyl-3,4-dimethoxy pyridine hydrochloride, 2-chloromethyl-3,4-dimethoxypyridinium chloride, 2-chloromethyl-3,4-dimethoxypyridinium hydrochloride, 2-chloromethyl-3 pound not4-dimethoxy-pyridine hydrochloride, 2-chloromethyl-3,4-dimethoxy pyridine hcl, 3,4-dimethoxy-2-chloromethylpyridine hydrochloride, 2-chloromethyl-3,4-dimethoxypyridine;hydrochloride, 2-chloromethyl-3,4-dimethoxy-pyridine hydrochloride
PubChem CID 16216928
IUPAC Name 2-(chloromethyl)-3,4-dimethoxypyridine;hydrochloride
SMILES COC1=C(C(=NC=C1)CCl)OC.Cl

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 5 g
UN Number 3077
Formula Weight 224.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Chloromethyl)-3,4-dimethoxypyridine Hydrochloride