accessibility menu, dialog, popup

UserName

H3CONClOCH3HCl

CAS RN 72830-09-2

IUPAC Name: 2-(chloromethyl)-3,4-dimethoxypyridine hydrochloride
Synonyms: 2-(chloromethyl)-3,4-dimethoxypyridine hydrochloride
Molecular Weight (g/mol): 224.08
Molecular Formula: C8H11Cl2NO2
InChi Key: YYRIKJFWBIEEDH-UHFFFAOYSA-N
SMILES: Cl.COC1=C(OC)C(CCl)=NC=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)