missing translation for 'onlineSavingsMsg'
Learn More

2-Chloromethyl-3,4-dimethoxypyridine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog No. p-7040459
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6023506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6023506 Supplier Thermo Scientific Chemicals Supplier No. H6023506

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 72830-09-2 | Molecular Formula: C8H11Cl2NO2 | Molecular Weight: 224.081 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 72830-09-2
Molecular Formula C8H11Cl2NO2
Molecular Weight (g/mol) 224.081
MDL Number MFCD02181083
InChI Key YYRIKJFWBIEEDH-UHFFFAOYSA-N
Synonym 2-chloromethyl-3,4-dimethoxypyridine hydrochloride, 2-chloromethyl-3,4-dimethoxy pyridine hydrochloride, 2-chloromethyl-3,4-dimethoxypyridinium chloride, 2-chloromethyl-3,4-dimethoxypyridinium hydrochloride, 2-chloromethyl-3 pound not4-dimethoxy-pyridine hydrochloride, 2-chloromethyl-3,4-dimethoxy pyridine hcl, 3,4-dimethoxy-2-chloromethylpyridine hydrochloride, 2-chloromethyl-3,4-dimethoxypyridine;hydrochloride, 2-chloromethyl-3,4-dimethoxy-pyridine hydrochloride
PubChem CID 16216928
IUPAC Name 2-(chloromethyl)-3,4-dimethoxypyridine;hydrochloride
SMILES COC1=C(C(=NC=C1)CCl)OC.Cl

Specifications

Melting Point 155°C (decomposition)
Quantity 5 g
UN Number UN3077
Formula Weight 224.08
Percent Purity 97%
Chemical Name or Material 2-Chloromethyl-3,4-dimethoxypyridine hydrochloride
Hazard Category H302+H312-H315-H317-H318-H373
Hazard Statement GHS H Statement
H318-H373-H302-H312-H317-H315
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Causes skin irritation.
Precautionary Statement P260-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P314-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 416-440-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only