missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethoxy)ethyltrimethylsilane (stabilized with Diisopropylethylamine) 95.0+%, TCI America™

Catalog No. p-7124923
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13395ML 5 mL
C133925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13395ML Supplier TCI America Supplier No. C13395ML
Only null left

Chemical Identifiers

CAS 76513-69-4
Molecular Formula C6H15ClOSi
Molecular Weight (g/mol) 166.72
MDL Number MFCD00009919
InChI Key BPXKZEMBEZGUAH-UHFFFAOYSA-N
Synonym 2-trimethylsilyl ethoxymethyl chloride, 2-chloromethoxy ethyl trimethylsilane, sem-chloride, 2-trimethylsilyl ethoxymethylchloride, sem-cl, 2-chloromethyl 2-trimethylsilyl ethyl ether, semcl, 2-chloromethoxy ethyl-trimethylsilane, 2-trimethysilyl-ethoxymethyl chloride
PubChem CID 2724271
IUPAC Name 2-(chloromethoxy)ethyl-trimethylsilane
SMILES C[Si](C)(C)CCOCCl

Specifications

Color Colorless
Boiling Point 59°C
Quantity 5 mL
UN Number 2920
Formula Weight 166.72
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Chloromethoxy)ethyltrimethylsilane (stabilized with Diisopropylethylamine)
TSCA No
Recommended Storage Refrigerator