missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroisonicotinaldehyde, 97%, Thermo Scientific™

Catalog No. p-7077597
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456040010 1g
AC456040050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456040010 Supplier Thermo Scientific Chemicals Supplier No. 456040010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 101066-61-9
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.55
MDL Number MFCD06651557
InChI Key UFPOSTQMFOYHJI-UHFFFAOYSA-N
Synonym 2-chloroisonicotinaldehyde, 2-chloro-4-pyridinecarboxaldehyde, 2-chloro-4-formylpyridine, 2-chloro-pyridine-4-carbaldehyde, 2-chloropyridine-4-carboxaldehyde, 4-pyridinecarboxaldehyde, 2-chloro, pubchem5099, 6-chloroisonicotinaldehyde, ksc180g9n, 2-chloropyridin-4-carbaldehyde
PubChem CID 2762994
IUPAC Name 2-chloropyridine-4-carbaldehyde
SMILES ClC1=CC(C=O)=CC=N1

Specifications

Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1g
Formula Weight 141.56
Percent Purity 97%
Chemical Name or Material 2-Chloroisonicotinaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapo

RUO – Research Use Only