accessibility menu, dialog, popup

UserName

ClON

CAS RN 101066-61-9

IUPAC Name: 2-chloropyridine-4-carbaldehyde
Synonyms: 2-chloropyridine-4-carbaldehyde
Molecular Weight (g/mol): 141.55
Molecular Formula: C6H4ClNO
InChi Key: UFPOSTQMFOYHJI-UHFFFAOYSA-N
SMILES: ClC1=CC(C=O)=CC=N1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)