missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroindole-3-carboxaldehyde, 97%

Catalog No. p-7032773
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3438103 1 g
AAH3438109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3438103 Supplier Thermo Scientific Chemicals Supplier No. H3438103

CAS: 5059-30-3 | C9H6ClNO | 179.603 g/mol

It is used as pharmaceutical intermediates. It is used in the synthesis of indole phytoalexin cyclobrassinon. And also in unprecedented chemical structure and biomimetic synthesis of erucalexin, a phytoalexin from the wild crucifer Erucastrum gallicum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates. It is used in the synthesis of indole phytoalexin cyclobrassinon. And also in unprecedented chemical structure and biomimetic synthesis of erucalexin, a phytoalexin from the wild crucifer Erucastrum gallicum.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store at room temperature.

Chemical Identifiers

CAS 5059-30-3
Molecular Formula C9H6ClNO
Molecular Weight (g/mol) 179.603
MDL Number MFCD00111620
InChI Key XYSSNBNFOBVMAU-UHFFFAOYSA-N
Synonym 2-chloro-indole-3-carbaldehyde, 2-chloroindole-3-carbaldehyde, 2-chloro-1h-indole-3-carboxaldehyde, 2-chloroindole-3-carboxaldehyde, 2-chloro-3-formyl-1h-indole, 1h-indole-3-carboxaldehyde, 2-chloro, chloroindolecarbaldehyde, 2-chloro-3-formylindol, 1h-indole-3-carboxaldehyde,2-chloro
PubChem CID 314791
IUPAC Name 2-chloro-1H-indole-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=C(N2)Cl)C=O

Specifications

Melting Point ∽212°C (decomposition)
Quantity 1 g
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 179.61
Percent Purity 97%
Chemical Name or Material 2-Chloroindole-3-carboxaldehyde
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only