accessibility menu, dialog, popup

UserName

ClHNO

CAS RN 5059-30-3

IUPAC Name: 2-chloro-1H-indole-3-carbaldehyde
Synonyms: 2-chloro-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 179.6
Molecular Formula: C9H6ClNO
InChi Key: XYSSNBNFOBVMAU-UHFFFAOYSA-N
SMILES: ClC1=C(C=O)C2=CC=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results