Learn More
2-Chloroimidazole, 97%
CAS: 16265-04-6 | C3H3ClN2 | 102.52 g/mol
Supplier: Thermo Scientific Chemicals H5186303
Description
2-Chloroimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloroimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
| 2-Chloroimidazole | |
| 165°C to 166°C | |
| MFCD02179530 | |
| 2-chloroimidazole, chloroimidazole, sftheadililuh@, pubchem7597, 2-chlor-1h-imidazol, zlchem 1064, acmc-209dos, 1h-imidazole,2-chloro, 2-chloranyl-1h-imidazole, 1h-imidazole, 2-chloro | |
| ClC1=NC=CN1 | |
| 102.52 | |
| CHEBI:52721 | |
| 97% |
| 16265-04-6 | |
| C3H3ClN2 | |
| 1 g | |
| OCVXSFKKWXMYPF-UHFFFAOYSA-N | |
| 2-chloro-1H-imidazole | |
| 2773328 | |
| 102.52 |
Chemical Identifiers
| 16265-04-6 | |
| 102.52 | |
| OCVXSFKKWXMYPF-UHFFFAOYSA-N | |
| 2773328 | |
| 2-chloro-1H-imidazole |
| C3H3ClN2 | |
| MFCD02179530 | |
| 2-chloroimidazole, chloroimidazole, sftheadililuh@, pubchem7597, 2-chlor-1h-imidazol, zlchem 1064, acmc-209dos, 1h-imidazole,2-chloro, 2-chloranyl-1h-imidazole, 1h-imidazole, 2-chloro | |
| CHEBI:52721 | |
| ClC1=NC=CN1 |
Safety and Handling
GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only