missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroimidazole, 97%

Catalog No. p-7034196
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51863MD 250 mg
AAH5186303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH51863MD Supplier Thermo Scientific Chemicals Supplier No. H51863MD

CAS: 16265-04-6 | C3H3ClN2 | 102.52 g/mol

2-Chloroimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 16265-04-6
Molecular Formula C3H3ClN2
Molecular Weight (g/mol) 102.52
MDL Number MFCD02179530
InChI Key OCVXSFKKWXMYPF-UHFFFAOYSA-N
Synonym 2-chloroimidazole, chloroimidazole, sftheadililuh@, pubchem7597, 2-chlor-1h-imidazol, zlchem 1064, acmc-209dos, 1h-imidazole,2-chloro, 2-chloranyl-1h-imidazole, 1h-imidazole, 2-chloro
PubChem CID 2773328
ChEBI CHEBI:52721
IUPAC Name 2-chloro-1H-imidazole
SMILES ClC1=NC=CN1

Specifications

Melting Point 165°C to 166°C
Quantity 250 mg
Formula Weight 102.52
Percent Purity 97%
Chemical Name or Material 2-Chloroimidazole
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only