missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroheptane 97.0+%, TCI America™
Supplier: TCI America C085225ML
Specifications
| 2-Chloroheptane | |
| Colorless | |
| C7H15Cl | |
| 25 mL | |
| 2-Heptyl Chloride | |
| CCCCCC(C)Cl | |
| 134.65 | |
| 134.65 | |
| Liquid |
| 1001-89-4 | |
| 146°C | |
| MFCD00039353 | |
| 1993 | |
| PTSLUOSUHFGQHV-UHFFFAOYNA-N | |
| 2-chloroheptane | |
| 34691 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 1001-89-4 | |
| 134.65 | |
| PTSLUOSUHFGQHV-UHFFFAOYNA-N | |
| 34691 | |
| CCCCCC(C)Cl |
| C7H15Cl | |
| MFCD00039353 | |
| 2-Heptyl Chloride | |
| 2-chloroheptane |
Safety and Handling
TSCA : No