missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroethyl Vinyl Ether (stabilized with MEHQ + Triethanolamine) 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | -70°C |
| Color | Yellow |
| Boiling Point | 108°C |
| Quantity | 25 mL |
| UN Number | 1992 |
| Formula Weight | 106.55 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Chloroethyl Vinyl Ether (stabilized with MEHQ + Triethanolamine) |
Safety and Handling
| EINECSNumber | (2)-0377 |
| RTECSNumber | KN6300000 |
| TSCA | Yes |