missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl Phenyl Sulfide 98.0+%, TCI America™

Catalog No. p-7124950
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13885G 5 g
C138825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13885G Supplier TCI America Supplier No. C13885G
Only null left

Chemical Identifiers

CAS 5535-49-9
Molecular Formula C8H9ClS
Molecular Weight (g/mol) 172.67
MDL Number MFCD00013691
InChI Key QDXIHHOPZFCEAP-UHFFFAOYSA-N
Synonym 2-chloroethyl phenyl sulfide, 2-chloroethyl phenyl sulfane, 2-chloroethyl sulfanyl benzene, 2-chloroethyl thio benzene, 2-chloroethylphenylsulfide, benzene, 2-chloroethyl thio, 2-phenylthio ethyl chloride, 2-phenylthioethylchloride, acmc-1axe6
PubChem CID 21702
IUPAC Name [(2-chloroethyl)sulfanyl]benzene
SMILES ClCCSC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 245°C
Quantity 5 g
UN Number 1760
Formula Weight 172.67
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Phenyl Sulfide