missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl Isothiocyanate 98.0+%, TCI America™

Catalog No. C22451G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C22451G 1 g
C22455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C22451G Supplier TCI America Supplier No. C22451G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6099-88-3
Molecular Formula C3H4ClNS
Molecular Weight (g/mol) 121.582
MDL Number MFCD00041125
InChI Key ZUWFBQUHBOUPFK-UHFFFAOYSA-N
Synonym 2-chloroethyl isothiocyanate, 2-chloroethylisothiocyanate, ethane, 1-chloro-2-isothiocyanato, sgxdlhbabrkth@, chloroethylisothiocyanate, acmc-1b2t4, beta-chloroethylisothiocyanate, beta-chloroethyl isothiocyanate, 1-chloro-2-isothiocyanato-ethane, #
PubChem CID 138661
IUPAC Name 1-chloro-2-isothiocyanatoethane
SMILES C(CCl)N=C=S

Specifications

Boiling Point 60°C
Quantity 1 g
UN Number 1992
Formula Weight 121.58
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Isothiocyanate