missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl isothiocyanate, 97%

Catalog No. AAL1160403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1160403 1 g
AAL1160406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1160403 Supplier Thermo Scientific Chemicals Supplier No. L1160403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6099-88-3 | C3H4ClNS | 121.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6099-88-3
Molecular Formula C3H4ClNS
Molecular Weight (g/mol) 121.582
MDL Number MFCD00041125
InChI Key ZUWFBQUHBOUPFK-UHFFFAOYSA-N
Synonym 2-chloroethyl isothiocyanate, 2-chloroethylisothiocyanate, ethane, 1-chloro-2-isothiocyanato, sgxdlhbabrkth@, chloroethylisothiocyanate, acmc-1b2t4, beta-chloroethylisothiocyanate, beta-chloroethyl isothiocyanate, 1-chloro-2-isothiocyanato-ethane, #
PubChem CID 138661
IUPAC Name 1-chloro-2-isothiocyanatoethane
SMILES C(CCl)N=C=S

Specifications

Density 1.265
Boiling Point 79°C to 80°C (13 mmHg)
Flash Point 96°C (205°F)
Refractive Index 1.556
Quantity 1 g
UN Number UN2922
Beilstein 605585
Sensitivity Moisture sensitive
Formula Weight 121.59
Percent Purity 97%
Chemical Name or Material 2-Chloroethyl isothiocyanate
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only