missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroethyl Ethyl Sulfide 98.0+%, TCI America™
$119.30 - $356.03
Chemical Identifiers
| CAS | 693-07-2 |
|---|---|
| Molecular Formula | C4H9ClS |
| Molecular Weight (g/mol) | 124.63 |
| MDL Number | MFCD00000979 |
| InChI Key | GBNVXYXIRHSYEG-UHFFFAOYSA-N |
| Synonym | 2-chloroethyl ethyl sulfide, 1-chloro-2-ethylthio ethane, ethane, 1-chloro-2-ethylthio, half-mustard gas, 2-ethylthioethyl chloride, 2-ethylthio chloroethane, sulfide, chloroethyl ethyl, ethyl 2-chloroethyl sulfide, h-mg, sulfide, 2-chloroethyl ethyl |
| PubChem CID | 12733 |
| IUPAC Name | 1-chloro-2-(ethylsulfanyl)ethane |
| SMILES | CCSCCCl |
Chemical Identifiers
| 693-07-2 | |
| 124.63 | |
| GBNVXYXIRHSYEG-UHFFFAOYSA-N | |
| 12733 | |
| CCSCCCl |
| C4H9ClS | |
| MFCD00000979 | |
| 2-chloroethyl ethyl sulfide, 1-chloro-2-ethylthio ethane, ethane, 1-chloro-2-ethylthio, half-mustard gas, 2-ethylthioethyl chloride, 2-ethylthio chloroethane, sulfide, chloroethyl ethyl, ethyl 2-chloroethyl sulfide, h-mg, sulfide, 2-chloroethyl ethyl | |
| 1-chloro-2-(ethylsulfanyl)ethane |
Specifications
| 693-07-2 | |
| 157°C | |
| MFCD00000979 | |
| 1992 | |
| GBNVXYXIRHSYEG-UHFFFAOYSA-N | |
| 1-chloro-2-(ethylsulfanyl)ethane | |
| 12733 | |
| ≥98.0% (GC) | |
| 2-Chloroethyl Ethyl Sulfide |
| Yellow | |
| C4H9ClS | |
| 5 mL | |
| 2-chloroethyl ethyl sulfide, 1-chloro-2-ethylthio ethane, ethane, 1-chloro-2-ethylthio, half-mustard gas, 2-ethylthioethyl chloride, 2-ethylthio chloroethane, sulfide, chloroethyl ethyl, ethyl 2-chloroethyl sulfide, h-mg, sulfide, 2-chloroethyl ethyl | |
| CCSCCCl | |
| 124.63 | |
| 124.63 | |
| Liquid |
Safety and Handling
RTECSNumber : WQ3250000
TSCA : Yes
Recommended Storage : Refrigerator