missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl Ethyl Ether 98.0+%, TCI America™

Catalog No. C117225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C117225ML 25 mL
C1172500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C117225ML Supplier TCI America Supplier No. C117225ML

Chemical Identifiers

CAS 628-34-2
Molecular Formula C4H9ClO
Molecular Weight (g/mol) 108.57
MDL Number MFCD00000974
InChI Key GPTVQTPMFOLLOA-UHFFFAOYSA-N
Synonym 2-chloroethyl ethyl ether, 2-ethoxyethyl chloride, 2-chloroethylethylether, 2-chloroethoxyethane, 1-chloro-2-ethoxy-ethane, ether, 2-chloroethyl ethyl, beta-chloroethyl ethyl ether, ethyl beta-chloroethyl ether, ethane, 1-chloro-2-ethoxy, ccris 9095
PubChem CID 12341
IUPAC Name 1-chloro-2-ethoxyethane
SMILES CCOCCCl

Specifications

Color Yellow
Boiling Point 108°C
Quantity 25 mL
UN Number 3271
Formula Weight 108.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Ethyl Ether
RTECSNumber KN6250000
TSCA No