accessibility menu, dialog, popup

UserName

H3COCl

CAS RN 628-34-2

IUPAC Name: 1-chloro-2-ethoxyethane
Synonyms: 2-chloroethylether
Molecular Weight (g/mol): 108.57
Molecular Formula: C4H9ClO
InChi Key: GPTVQTPMFOLLOA-UHFFFAOYSA-N
SMILES: CCOCCCl
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)