missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl Chloroformate 98.0+%, TCI America™

Catalog No. p-7124762
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C105225G 25 g
C1052100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C105225G Supplier TCI America Supplier No. C105225G

Chemical Identifiers

CAS 627-11-2
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.96
MDL Number MFCD00000646
InChI Key SVDDJQGVOFZBNX-UHFFFAOYSA-N
Synonym 2-chloroethyl chloroformate, chloroethyl chloroformate, carbonochloridic acid, 2-chloroethyl ester, chloroformic acid 2-chloroethyl ester, formic acid, chloro-, 2-chloroethyl ester, chloroethylchloroformate, 2-chloroethylchloroformate, unii-4d32h07z9i, beta-chloroethyl chlorocarbonate, chloroformic acid, beta-chloroethyl ester
PubChem CID 12305
IUPAC Name 2-chloroethyl carbonochloridate
SMILES ClCCOC(Cl)=O

Specifications

Color Yellow
Boiling Point 154°C
Quantity 25 g
UN Number 2742
Formula Weight 142.96
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl Chloroformate
EINECSNumber (2)-3629
RTECSNumber LQ5950000
TSCA Yes
Recommended Storage Refrigerator