accessibility menu, dialog, popup

UserName

ClOClO

CAS RN 627-11-2

IUPAC Name: 2-chloroethyl carbonochloridate
Synonyms: chloroethyl chloroformate
Molecular Weight (g/mol): 142.96
Molecular Formula: C3H4Cl2O2
InChi Key: SVDDJQGVOFZBNX-UHFFFAOYSA-N
SMILES: ClCCOC(Cl)=O
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)